MAN326368480 Manchester Organics

2-Bromo-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD06208253
  • Molecular Weight : 269.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P22272

Safety Information

MSDS https://www.manchesterorganics.com/P22272
Availability details for
Quantity
  • Availability
  • Add to Cart