MAN326368481 Manchester Organics

4-Fluorophenylacetic acid ethyl ester

  • MDL number MFCD08235152
  • Molecular Weight : 182.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U23460

Safety Information

MSDS https://www.manchesterorganics.com/U23460
Availability details for
Quantity
  • Availability
  • Add to Cart