MAN326368495 Manchester Organics

1-Amino-2-naphthalenecarboxylic acid

  • MDL number MFCD11193636
  • Cas Number : 4919-43-1
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-amino-2-naphthoic acid
Inchi Code 1S/C11H9NO2/c12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h1-6H,12H2,(H,13,14)
Inchi Key VFFRLRQQWXGEBX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/B23135
Availability details for
Quantity
  • Availability
  • Add to Cart