MAN326368526 Manchester Organics

2-Amino-4-chlorobenzoic acid methyl ester

  • MDL number MFCD00017568
  • Molecular Weight : 185.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K23216

Safety Information

MSDS https://www.manchesterorganics.com/K23216
Availability details for
Quantity
  • Availability
  • Add to Cart