MAN326368637 Manchester Organics

4-Methoxy-3-pyridineboronic acid pinacol ester

  • MDL number MFCD07367561
  • Molecular Weight : 235.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B23208

Safety Information

MSDS https://www.manchesterorganics.com/B23208
Availability details for
Quantity
  • Availability
  • Add to Cart