MAN326368640 Manchester Organics

2-Chlorophenylacetone

  • MDL number MFCD00045175
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D23281

Safety Information

MSDS https://www.manchesterorganics.com/D23281
Availability details for
Quantity
  • Availability
  • Add to Cart