MAN326368641 Manchester Organics

3-Chlorophenylacetone

  • MDL number MFCD00082872
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E23257

Safety Information

MSDS https://www.manchesterorganics.com/E23257
Availability details for
Quantity
  • Availability
  • Add to Cart