MAN326368642 Manchester Organics

4-Chlorophenylacetone

  • MDL number MFCD00045214
  • Cas Number : 5586-88-9
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/F23187
Availability details for
Quantity
  • Availability
  • Add to Cart