MAN326368643 Manchester Organics

2,5-Dichloropyridine-4-boronic acid pinacol ester

  • MDL number MFCD06798256
  • Molecular Weight : 273.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H23260

Safety Information

MSDS https://www.manchesterorganics.com/H23260
Availability details for
Quantity
  • Availability
  • Add to Cart