MAN326368678 Manchester Organics

1-(2-Trimethylsilanylethoxymethyl)-1H-pyrazole-5-boronic acid pinacol ester

  • MDL number MFCD09037502
  • Molecular Weight : 324.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J23215
Availability details for
Quantity
  • Availability
  • Add to Cart