MAN326368687 Manchester Organics

4-Bromo-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD09037472
  • Cas Number : 861928-27-0
  • Molecular Weight : 253.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/S23222
Availability details for
Quantity
  • Availability
  • Add to Cart