MAN326368694 Manchester Organics

2-Bromo-5-methylpyrazine

  • MDL number MFCD08705754
  • Molecular Weight : 173.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y23228
Availability details for
Quantity
  • Availability
  • Add to Cart