MAN326368697 Manchester Organics

5-Methyl-1,3,4-oxadiazol-2-ylamine

  • MDL number MFCD03703470
  • Cas Number : 52838-39-8
  • Molecular Weight : 99.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1,3,4-oxadiazol-2-amine
Inchi Code 1S/C3H5N3O/c1-2-5-6-3(4)7-2/h1H3,(H2,4,6)
Inchi Key XPXWYVCQCNFIIJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R23221
Availability details for
Quantity
  • Availability
  • Add to Cart