MAN326368702 Manchester Organics

5-Amino-2-methyl benzoic acid methyl ester

  • MDL number MFCD08752568
  • Molecular Weight : 165.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F23306

Safety Information

MSDS https://www.manchesterorganics.com/F23306
Availability details for
Quantity
  • Availability
  • Add to Cart