MAN326368714 Manchester Organics

4-Chloro-5-methylpyridine-2-carboxylic acid

  • MDL number MFCD12400883
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V23603

Safety Information

MSDS https://www.manchesterorganics.com/V23603
Availability details for
Quantity
  • Availability
  • Add to Cart