MAN326368715 Manchester Organics

2,3-Difluoropyridine-6-carboxylic acid

  • MDL number MFCD09031175
  • Molecular Weight : 159.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U23579

Safety Information

MSDS https://www.manchesterorganics.com/U23579
Availability details for
Quantity
  • Availability
  • Add to Cart