MAN326368716 Manchester Organics

4,4-Dimethylcyclohexylamine

  • MDL number MFCD08460046
  • Molecular Weight : 127.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y23725

Safety Information

MSDS https://www.manchesterorganics.com/Y23725
Availability details for
Quantity
  • Availability
  • Add to Cart