MAN326368753 Manchester Organics

2-Bromo-4-nitropyridine-N-oxide

  • MDL number MFCD00160743
  • Molecular Weight : 218.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B23302

Safety Information

MSDS https://www.manchesterorganics.com/B23302
Availability details for
Quantity
  • Availability
  • Add to Cart