MAN326368758 Manchester Organics

2-Bromo-5-methoxyphenol

  • MDL number MFCD03428523
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S12263

Safety Information

MSDS https://www.manchesterorganics.com/S12263
Availability details for
Quantity
  • Availability
  • Add to Cart