MAN326368761 Manchester Organics

3-(7-Ethylindole)ethanol

  • MDL number MFCD01718805
  • Molecular Weight : 189.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D23352

Safety Information

MSDS https://www.manchesterorganics.com/D23352
Availability details for
Quantity
  • Availability
  • Add to Cart