MAN326368777 Manchester Organics

2,4-Dichloro-5-pyrimidine carboxylic acid ethyl ester

  • MDL number MFCD09910281
  • Cas Number : 51940-64-8
  • Molecular Weight : 221.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2,4-dichloro-5-pyrimidinecarboxylate
Inchi Code 1S/C7H6Cl2N2O2/c1-2-13-6(12)4-3-10-7(9)11-5(4)8/h3H,2H2,1H3
Inchi Key SRJBDGLSCPDXBL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/K23287
Availability details for
Quantity
  • Availability
  • Add to Cart