MAN326368783 Manchester Organics

(4-Chloro-pyridin-2-yl)-dimethyl-amine

  • MDL number MFCD09031036
  • Molecular Weight : 156.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S23481

Safety Information

MSDS https://www.manchesterorganics.com/S23481
Availability details for
Quantity
  • Availability
  • Add to Cart