MAN326368915 Manchester Organics

Docosanal

  • MDL number MFCD07780595
  • Molecular Weight : 324.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y12081
Availability details for
Quantity
  • Availability
  • Add to Cart