MAN326369071 Manchester Organics

Methyl 2-(2-chloroethoxy)acetate

  • MDL number MFCD09030969
  • Cas Number : 83881-47-4
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J23499
Availability details for
Quantity
  • Availability
  • Add to Cart