MAN326369114 Manchester Organics

2-(1-(tert-Butoxycarbonyl)azetidin-3-yl)acetic acid

  • MDL number MFCD02179028
  • Cas Number : 183062-96-6
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U22418
Availability details for
Quantity
  • Availability
  • Add to Cart