MAN326369139 Manchester Organics

3-Hydroxy-5-methylbenzoic acid

  • MDL number MFCD09833213
  • Molecular Weight : 152.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F23638
Availability details for
Quantity
  • Availability
  • Add to Cart