MAN326369143 Manchester Organics

1-(Boc-amino)-3-(methylamino)propane

  • MDL number MFCD06808586
  • Cas Number : 442514-22-9
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C22332
Availability details for
Quantity
  • Availability
  • Add to Cart