MAN326369240 Manchester Organics

2,4-Dichlorothieno[3,2-d]pyrimidine

  • MDL number MFCD08448158
  • Molecular Weight : 205.064

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V23177
Availability details for
Quantity
  • Availability
  • Add to Cart