MAN326369248 Manchester Organics

4-Methoxyphenoxyacetic acid

  • MDL number MFCD00014360
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/U23721
Availability details for
Quantity
  • Availability
  • Add to Cart