MAN326369274 Manchester Organics

1-Isopropyl-4-piperidinol

  • MDL number MFCD00101977
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z23939
Availability details for
Quantity
  • Availability
  • Add to Cart