MAN326369348 Manchester Organics

Methyl 3-amino-5-(trifluoromethyl)benzoate

  • MDL number MFCD00625827
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M13821

Safety Information

MSDS https://www.manchesterorganics.com/M13821
Availability details for
Quantity
  • Availability
  • Add to Cart