MAN326369350 Manchester Organics

3-Bromo-4-(difluoromethyl)pyridine

  • MDL number MFCD18257232
  • Molecular Weight : 208.00346

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H12090

Safety Information

MSDS https://www.manchesterorganics.com/H12090
Availability details for
Quantity
  • Availability
  • Add to Cart