MAN372903013 Manchester Organics

(2-Amino-1,1-dimethyl-ethyl)-carbamic acid tert-butyl ester

  • MDL number MFCD14529625
  • Molecular Weight : 188.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W24575

Safety Information

MSDS https://www.manchesterorganics.com/W24575
Availability details for
Quantity
  • Availability
  • Add to Cart