MAN372903020 Manchester Organics

2-Fluoro-4-methylpyridine-5-boronic acid

  • MDL number MFCD08741406
  • Molecular Weight : 154.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X20387

Safety Information

MSDS https://www.manchesterorganics.com/X20387
Availability details for
Quantity
  • Availability
  • Add to Cart