MAN372903028 Manchester Organics

5-Chloro-2-fluoropyridine-4-boronic acid

  • MDL number MFCD05662405
  • Cas Number : 1034659-38-5
  • Molecular Weight : 175.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/U21921
Availability details for
Quantity
  • Availability
  • Add to Cart