MAN372903056 Manchester Organics

1-(2-Amino-5-chlorophenyl-2,2,2-trifluoroethanone

  • MDL number MFCD08458216
  • Cas Number : 154598-53-5
  • Molecular Weight : 223.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C24226
Availability details for
Quantity
  • Availability
  • Add to Cart