MAN372903060 Manchester Organics

2-Chloro-5-(trifluoromethoxy)pyridine

  • MDL number MFCD18257007
  • Cas Number : 1206972-45-3
  • Molecular Weight : 197.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y12104
Availability details for
Quantity
  • Availability
  • Add to Cart