MAN372903071 Manchester Organics

(6-Fluoro-3-pyridinyl)methanol

  • MDL number MFCD03095186
  • Cas Number : 39891-05-9
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/K12115
Availability details for
Quantity
  • Availability
  • Add to Cart