MAN372903128 Manchester Organics

1-Benzyloxy-2-nitro-benzene

  • MDL number MFCD00143680
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P12119
Availability details for
Quantity
  • Availability
  • Add to Cart