MAN372903233 Manchester Organics

(5-Fluoro-3-pyridyl)methanol

  • MDL number MFCD01758680
  • Cas Number : 22620-32-2
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B12130
Availability details for
Quantity
  • Availability
  • Add to Cart