MAN372903252 Manchester Organics

(6-Fluoro-2-pyridyl)methanol

  • MDL number MFCD07783827
  • Cas Number : 315180-17-7
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-fluoro-2-pyridinyl)methanol
Inchi Code 1S/C6H6FNO/c7-6-3-1-2-5(4-9)8-6/h1-3,9H,4H2
Inchi Key OWTCZNAMUJIMOC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T12147
Availability details for
Quantity
  • Availability
  • Add to Cart