MAN372903267 Manchester Organics

6-Bromo-7-azaindole

  • MDL number MFCD06659667
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J24468
Availability details for
Quantity
  • Availability
  • Add to Cart