MAN372903311 Manchester Organics

5-Hydroxy-2-methyl benzoic acid methyl ester

  • MDL number MFCD11226199
  • Cas Number : 73505-48-3
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A24531
Availability details for
Quantity
  • Availability
  • Add to Cart