MAN372903322 Manchester Organics

5-Amino-3-isoxazole carboxylic acid ethyl ester

  • MDL number MFCD13174972
  • Molecular Weight : 156.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E23518
Availability details for
Quantity
  • Availability
  • Add to Cart