MAN372903451 Manchester Organics

2-Cyano-3,6-dimethylpyridine

  • MDL number MFCD06637568
  • Cas Number : 68164-77-2
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/E12158
Availability details for
Quantity
  • Availability
  • Add to Cart