MAN372903609 Manchester Organics

2,5-Dihydroxythiophenol

  • MDL number MFCD00186219
  • Molecular Weight : 142.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N25136

Safety Information

MSDS https://www.manchesterorganics.com/N25136
Availability details for
Quantity
  • Availability
  • Add to Cart