MAN372903842 Manchester Organics

5-Chloro-2-methylpyridine

  • MDL number MFCD04114188
  • Cas Number : 72093-07-3
  • Molecular Weight : 127.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F25033
Availability details for
Quantity
  • Availability
  • Add to Cart