MAN372903906 Manchester Organics

5-Bromo-3-chloropyridazine

  • MDL number MFCD13190293
  • Cas Number : 1196155-33-5
  • Molecular Weight : 193.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/A25051
Availability details for
Quantity
  • Availability
  • Add to Cart