MAN372903907 Manchester Organics

1-Methyl-1H-benzimidazole-2-carboxylic acid

  • MDL number MFCD01836869
  • Cas Number : 20572-01-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B25052
Availability details for
Quantity
  • Availability
  • Add to Cart