MAN372903930 Manchester Organics

4-Amino-5,6-dichloropyrimidine

  • MDL number MFCD09837293
  • Cas Number : 310400-38-5
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N23810
Availability details for
Quantity
  • Availability
  • Add to Cart